Assay Details
Gene Symbol: MOB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618863.1 | XP_023474631.1 | 790 | 71 | ||
XM_023618865.1 | XP_023474633.1 | 577 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
