Assay Details
Gene Symbol: TPRKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488244.6 | XP_001488294.3 | 348 | 77 | ||
XM_003362970.4 | XP_003363018.1 | 301 | 77 | |||
XM_014731091.2 | XP_014586577.1 | 529 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
