Assay Details
Gene Symbol: ALMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618885.1 | XP_023474653.1 | 8918 | 64 | ||
GenBank mRNA | JL633258.1 | - | 1 - 2 | 267 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
