Assay Details
Gene Symbol: TGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599909.2 | XP_005599966.1 | 643 | 68 | ||
XM_005599910.2 | XP_005599967.1 | 638 | 68 | |||
XM_023618943.1 | XP_023474711.1 | 1646 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
