Assay Details
Gene Symbol: NFU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491049.6 | XP_001491099.1 | 544 | 89 | ||
XM_023618964.1 | XP_023474732.1 | 457 | 89 | |||
XM_023618965.1 | XP_023474733.1 | 283 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
