Assay Details
Gene Symbol: CNRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492209.5 | XP_001492259.1 | 716 | 74 | ||
XM_014731125.2 | XP_014586611.1 | 697 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
