Assay Details
Gene Symbol: C1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492549.4 | XP_001492599.2 | 329 | 105 | ||
XM_005599928.3 | XP_005599985.1 | 240 | 105 | |||
GenBank mRNA | JL637382.1 | - | 2 - 3 | 176 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
