Assay Details
Gene Symbol: COMMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494799.5 | XP_001494849.1 | 498 | 89 | ||
XM_023619035.1 | XP_023474803.1 | 498 | 89 | |||
XM_023619036.1 | XP_023474804.1 | 498 | 89 | |||
XM_023619037.1 | XP_023474805.1 | 762 | 89 | |||
XM_023619038.1 | XP_023474806.1 | 520 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
