Assay Details
Gene Symbol: FOXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619116.1 | XP_023474884.1 | 855 | 76 | ||
XM_023619117.1 | XP_023474885.1 | 786 | 76 | |||
XM_023619118.1 | XP_023474886.1 | 864 | 76 | |||
XM_023619119.1 | XP_023474887.1 | 753 | 76 | |||
XM_023619120.1 | XP_023474888.1 | 1047 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
