Assay Details
Gene Symbol: CRIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362995.4 | XP_003363043.1 | 369 | 106 | ||
GenBank mRNA | JL615994.1 | - | 4 - 5 | 328 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
