Assay Details
Gene Symbol: CDKL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500269.6 | XP_001500319.4 | 638 | 64 | ||
XM_014731191.2 | XP_014586677.1 | 638 | 64 | |||
XM_023619176.1 | XP_023474944.1 | 638 | 64 | |||
XM_023619177.1 | XP_023474945.1 | 633 | 64 | |||
XM_023619178.1 | XP_023474946.1 | 559 | 64 | |||
XM_023619179.1 | XP_023474947.1 | 638 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
