Assay Details
Gene Symbol: DPY30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501494.4 | XP_001501544.1 | 222 | 79 | ||
XM_001501502.5 | XP_001501552.1 | 161 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
