Assay Details
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500475.5 | XP_001500525.3 | 665 | 72 | ||
XM_005600096.3 | XP_005600153.2 | 495 | 72 | |||
XM_014731229.2 | XP_014586715.2 | 686 | 72 | |||
XM_023619295.1 | XP_023475063.1 | 599 | 72 | |||
XM_023619296.1 | XP_023475064.1 | 324 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
