Assay Details
Gene Symbol: LOC100070891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618435.1 | XP_023474203.1 | 2001 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
