Assay Details
Gene Symbol: BABAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502017.5 | XP_001502067.1 | 133 | 87 | ||
GenBank mRNA | JL616309.1 | - | 1 - 2 | 62 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502017.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
