Assay Details
Gene Symbol: BABAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502017.5 | XP_001502067.1 | 937 | 94 | ||
XM_023619317.1 | XP_023475085.1 | 828 | 94 | |||
GenBank mRNA | JL616309.1 | - | 8 - 9 | 866 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
