Assay Details
Gene Symbol: ATRAID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600129.3 | XP_005600186.1 | 256 | 66 | ||
GenBank mRNA | JL626206.1 | - | 5 - 6 | 613 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
