Assay Details
Gene Symbol: ASXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001918196.4 | XP_001918231.2 | 910 | 64 | ||
XM_005600159.3 | XP_005600216.2 | 907 | 64 | |||
XM_005600160.3 | XP_005600217.2 | 266 | 64 | |||
XM_005600161.3 | XP_005600218.2 | 2622 | 64 | |||
XM_005600162.3 | XP_005600219.2 | 1098 | 64 | |||
XM_023619373.1 | XP_023475141.1 | 727 | 64 | |||
XM_023619374.1 | XP_023475142.1 | 637 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
