Assay Details
Gene Symbol: POMC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014731265.2 | XP_014586751.2 | 310 | 64 | ||
XM_014731267.2 | XP_014586753.2 | 364 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
