Assay Details
Gene Symbol: TP53I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619425.1 | XP_023475193.1 | 513 | 69 | ||
XM_023619426.1 | XP_023475194.1 | 513 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
