Assay Details
Gene Symbol: CYRIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600202.3 | XP_005600259.1 | 1760 | 65 | ||
XM_005600208.3 | XP_005600265.1 | 3948 | 65 | |||
XM_014731299.2 | XP_014586785.1 | 853 | 65 | |||
XM_023619492.1 | XP_023475260.1 | 864 | 65 | |||
XM_023619493.1 | XP_023475261.1 | 1649 | 65 | |||
XM_023619494.1 | XP_023475262.1 | 1788 | 65 | |||
XM_023619496.1 | XP_023475264.1 | 1629 | 65 | |||
XM_023619497.1 | XP_023475265.1 | 1384 | 65 | |||
XM_023619498.1 | XP_023475266.1 | 1238 | 65 | |||
XM_023619499.1 | XP_023475267.1 | 1008 | 65 | |||
XM_023619500.1 | XP_023475268.1 | 1010 | 65 | |||
XM_023619501.1 | XP_023475269.1 | 641 | 65 | |||
GenBank mRNA | JL615922.1 | - | 6 - 7 | 641 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
