Assay Details
Gene Symbol: NBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014731302.2 | XP_014586788.1 | 2614 | 68 | ||
XM_023619505.1 | XP_023475273.1 | 2575 | 68 | |||
XM_023619506.1 | XP_023475274.1 | 2614 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
