Assay Details
Gene Symbol: SLC66A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502263.5 | XP_001502313.3 | 497 | 100 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC66A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502263.5 | XP_001502313.3 | 497 | 100 |