Assay Details
Gene Symbol: RRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014731312.2 | XP_014586798.2 | 508 | 74 | ||
XM_023619542.1 | XP_023475310.1 | 513 | 74 | |||
XM_023619543.1 | XP_023475311.1 | 513 | 74 | |||
XM_023619544.1 | XP_023475312.1 | 513 | 74 | |||
XM_023619545.1 | XP_023475313.1 | 513 | 74 | |||
XM_023619546.1 | XP_023475314.1 | 513 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
