Assay Details
Gene Symbol: IAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619557.1 | XP_023475325.1 | 495 | 63 | ||
XM_023619558.1 | XP_023475326.1 | 448 | 63 | |||
XM_023619559.1 | XP_023475327.1 | 438 | 63 | |||
XM_023619560.1 | XP_023475328.1 | 325 | 63 | |||
XM_023619561.1 | XP_023475329.1 | 411 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
