Assay Details
Gene Symbol: CMPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619592.1 | XP_023475360.1 | 1086 | 103 | ||
XM_023619593.1 | XP_023475361.1 | 1086 | 103 |

More Information
Set Membership: |
![]() ![]() |
