Assay Details
Gene Symbol: ACP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503658.5 | XP_001503708.1 | 142 | 73 | ||
XM_001503660.6 | XP_001503710.1 | 142 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
