Assay Details
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619873.1 | XP_023475641.1 | 619 | 81 | ||
GenBank mRNA | JL639024.1 | - | 1 - 2 | 217 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
