Assay Details
Gene Symbol: CNBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363052.4 | XP_003363100.1 | 571 | 68 | ||
XM_003363053.4 | XP_003363101.1 | 586 | 68 | |||
XM_005600337.3 | XP_005600394.1 | 589 | 68 | |||
XM_005600338.3 | XP_005600395.1 | 568 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
