Assay Details
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619901.1 | XP_023475669.1 | 638 | 64 | ||
XM_023619902.1 | XP_023475670.1 | 639 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
