Assay Details
Gene Symbol: MKRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023619945.1 | XP_023475713.1 | 1129 | 66 | ||
XM_023619946.1 | XP_023475714.1 | 1023 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
