Assay Details
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494697.5 | XP_001494747.1 | 1145 | 60 | ||
XM_005600384.3 | XP_005600441.1 | 1128 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
