Assay Details
Gene Symbol: EIF4E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621182.1 | XP_023476950.1 | 421 | 77 | ||
XM_023621183.1 | XP_023476951.1 | 421 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
