Assay Details
Gene Symbol: PRICKLE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600491.3 | XP_005600548.1 | 2238 | 55 | ||
XM_005600492.3 | XP_005600549.1 | 6472 | 55 | |||
XM_023620119.1 | XP_023475887.1 | 2736 | 55 | |||
XM_023620120.1 | XP_023475888.1 | 2050 | 55 | |||
XM_023620121.1 | XP_023475889.1 | 2039 | 55 | |||
XM_023620122.1 | XP_023475890.1 | 2084 | 55 | |||
XM_023620123.1 | XP_023475891.1 | 1930 | 55 | |||
XM_023620124.1 | XP_023475892.1 | 6476 | 55 | |||
XM_023620126.1 | XP_023475894.1 | 1880 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
