Assay Details
Gene Symbol: ABHD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600595.3 | XP_005600652.1 | 836 | 87 | ||
XM_014731634.2 | XP_014587120.1 | 938 | 87 | |||
XM_014731635.2 | XP_014587121.1 | 921 | 87 | |||
XM_014731636.2 | XP_014587122.1 | 916 | 87 | |||
XM_014731637.2 | XP_014587123.1 | 1052 | 87 | |||
XM_014731638.2 | XP_014587124.1 | 832 | 87 | |||
XM_014731639.2 | XP_014587125.1 | 890 | 87 | |||
XM_023620351.1 | XP_023476119.1 | 829 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
