Assay Details
Gene Symbol: CISH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493420.5 | XP_001493470.2 | 163 | 56 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001493420.5 transcripts.
Set Membership: |
![]() ![]() |
