Assay Details
Gene Symbol: RASSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014731655.2 | XP_014587141.2 | 612 | 84 | ||
XM_023620385.1 | XP_023476153.1 | 831 | 84 | |||
XM_023620386.1 | XP_023476154.1 | 741 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
