Assay Details
Gene Symbol: HYAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600628.3 | XP_005600685.1 | 1515 | 69 | ||
XM_005600629.3 | XP_005600686.1 | 1113 | 69 | |||
XM_005600630.3 | XP_005600687.1 | 1299 | 69 | |||
XM_023620376.1 | XP_023476144.1 | 1799 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
