Assay Details
Gene Symbol: IP6K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497160.6 | XP_001497210.1 | 715 | 60 | ||
XM_005600655.3 | XP_005600712.1 | 1041 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
