Assay Details
Gene Symbol: QARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498296.6 | XP_001498346.1 | 1654 | 58 | ||
XM_005600682.3 | XP_005600739.1 | 1562 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
