Assay Details
Gene Symbol: ABHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023620624.1 | XP_023476392.1 | 757 | 64 | ||
XM_023620625.1 | XP_023476393.1 | 670 | 64 | |||
XM_023620626.1 | XP_023476394.1 | 568 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
