Assay Details
Gene Symbol: SNRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023620628.1 | XP_023476396.1 | 835 | 65 | ||
XM_023620629.1 | XP_023476397.1 | 900 | 65 | |||
XM_023620630.1 | XP_023476398.1 | 884 | 65 | |||
XM_023620631.1 | XP_023476399.1 | 789 | 65 | |||
XM_023620632.1 | XP_023476400.1 | 805 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
