Assay Details
Gene Symbol: DLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600820.3 | XP_005600877.2 | 4312 | 59 | ||
XM_014731783.2 | XP_014587269.2 | 4312 | 59 | |||
XM_023620735.1 | XP_023476503.1 | 4219 | 59 | |||
XM_023620736.1 | XP_023476504.1 | 4001 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
