Assay Details
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023620792.1 | XP_023476560.1 | 490 | 72 | ||
XM_023620793.1 | XP_023476561.1 | 361 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
