Assay Details
Gene Symbol: CMTM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023620794.1 | XP_023476562.1 | 544 | 79 | ||
XM_023620795.1 | XP_023476563.1 | 529 | 79 | |||
XM_023620798.1 | XP_023476566.1 | 2708 | 79 | |||
XM_023620799.1 | XP_023476567.1 | 561 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
