Assay Details
Gene Symbol: METTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496483.5 | XP_001496533.1 | 561 | 99 | ||
XM_005600924.3 | XP_005600981.1 | 561 | 99 | |||
XM_023620908.1 | XP_023476676.1 | 561 | 99 | |||
XM_023620909.1 | XP_023476677.1 | 624 | 99 | |||
XM_023620910.1 | XP_023476678.1 | 546 | 99 | |||
XM_023620912.1 | XP_023476680.1 | 561 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
