Assay Details
Gene Symbol: IL20RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014731885.2 | XP_014587371.1 | 632 | 73 | ||
XM_014731886.2 | XP_014587372.1 | 636 | 73 | |||
XM_014731887.2 | XP_014587373.1 | 633 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
