Assay Details
Gene Symbol: FAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600954.3 | XP_005601011.2 | 543 | 68 | ||
XM_023621012.1 | XP_023476780.1 | 548 | 68 | |||
XM_023621013.1 | XP_023476781.1 | 623 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
