Assay Details
Gene Symbol: TSC22D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621101.1 | XP_023476869.1 | 2260 | 99 | ||
XM_023621102.1 | XP_023476870.1 | 2239 | 99 | |||
XM_023621103.1 | XP_023476871.1 | 2159 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
