Assay Details
Gene Symbol: SLC33A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014731544.2 | XP_014587030.2 | 1893 | 66 | ||
XM_023621171.1 | XP_023476939.1 | 1893 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
